1018001
USP
N-Acetyl-L-leucine
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Acetyl-L-leucine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
API family
leucine
form
solid
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
It is intended to be used as a USP reference standard in specified quality tests and assays. It is also used to prepare standard solutions in the system suitability testing of liquid chromatographic system as per USP monograph citrulline.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Citruline
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(4), 4908-4908 (2020)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service