Key Documents
Z227323
Supelco® Precolumn Filter
Direct-connect, PEEK
Synonym(s):
TM=["Supelco"] Guard Column Filter
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
material
PEEK
description
starter kit
parameter
100 °C max. temp.
5000 psi max. pressure
bore
0.015 in.
General description
Direct-connect; protects analytical and guard columns. Our precolumn filter can be connected directly, hand-tightened, into any HPLC column or guard column that has Valco-compatible end fittings. PEEK cap and body, 2 μm stainless steel frit. For a metal-free system, order PEEK (biocompatible) replacement frits (57430-U).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
recommended
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Questions
  1. Hi l want to know more information about this Suprlcp Precolumn Filter, is this the plastic one or steel one? Is this the only filter or it is the whole precolumn within the filter? Could you please send me a picture about this product?
    1 answer
    1. This product is made of polyetheretherketone (PEEK), which is a high-performance plastic. The filter is secured between the two end pieces.
      Images are provided below.
      See the links below to review the compatible stainless steel frits;
      Z227331, pore size 2.0 ?m
      https://www.sigmaaldrich.com/product/supelco/z227331
      Z290874, pore size 0.5 ?m
      https://www.sigmaaldrich.com/product/supelco/z290874
      Helpful?