Skip to Content
Merck
All Photos(1)

Key Documents

807457

Supelco

TOYOPEARL® HW-55F Bulk Media

phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size

Synonym(s):

TOYOPEARL® HW-55 Size Exclusion Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

description

HW-55F

form

beads

packaging

bottle of 500 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable
LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

45 μm

pore size

1000-200,000 Da MW range

operating pH

2-13

separation technique

size exclusion (SEC)

General description

Toyopearl HW-55 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 1,000-700,000 Da. It also is the base bead for all Toyopearl "-550" designated products.

Application

TOYOPEARL® media fractionate mixtures of proteins and other large molecular weight compounds over a wide size range. Applications include separating RNA from protein, resolving oligosaccharides by degree of polymerization, and isolating agglutinin while maintaining a high hemagglutination titer.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga N Smol'kina et al.
Carbohydrate research, 347(1), 161-163 (2011-11-22)
A method is developed for the preparation of D-rhamnose from an O-polysaccharide (OPS) isolated by mild acid hydrolysis of Azospirillum brasilense SR75 cell mass. After the OPS hydrolysis, D-rhamnose was recovered by gel-permeation chromatography on Toyopearl TSK HW-40 and was
M V Pokrovskaya et al.
Protein expression and purification, 82(1), 150-154 (2012-01-10)
We have cloned ansB (YPTB1411) gene from Yersinia pseudotuberculosis Q66CJ2 and constructed stable inducible expression system that overproduce L-asparaginase from Y. pseudotuberculosis (YpA) in Escherichiacoli BL21 (DE3) cells. For purification of YpA we used Q-Sepharose and DEAE-Toyopearl column chromatography. We
Shu-fen Xu et al.
Fitoterapia, 83(1), 153-160 (2011-11-01)
Persimmon proanthocyanidin was fractionated on Toyopearl TSK-HW-50-F to yield a fraction with strong inhibition on the catalytic activity and edema-inducing activity and lethality of Chinese cobra PLA(2). Thiolysis suggested that the terminal units included C, EGCG and myricetin, and epicatechin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service