59686-U
Supel™ BioSPME 96-Pin Device Positioning Adapter
Synonym(s):
Supel™ BioSPME 96-Pin Device Positioning Adapter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 1 ea
Quality Level
application(s)
clinical
pharmaceutical
compatibility
for use with (Supel™ BioSPME 96-Pin Device)
for use with (automated robotic liquid handling systems)
General description
The Supel™ BioSPME 96-Pin Device Positioning Adapter is used to securely and accurately position the Supel™ BioSPME 96-Pin Device into the robotic deck during the automated solid phase microextraction (SPME) workflow. The adapter should be situated in a position on the automation deck where the BioSPME device will rest while not in use (i.e. before the extraction workflow begins, after the extraction workflow is completed, and/or during steps when the grippers are needed to perform an alternative action).
Recommended products
Supel™ BioSPME 96-Pin Devices 59680-U and 59683-U
Legal Information
Supel is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service