509353
SGE Syringe Valves
Push button with Luer Lock, for use with SGE 5mL-100mL capacity syringe, volume 5-2000 mL
Synonym(s):
SYRINGE VALVE, LUER LOCK, 5ML-100ML
About This Item
Recommended Products
material
stainless steel body
Agency
suitable for EPA 601
feature
Push Button with Luer Lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 031910
volume
5-2000 mL
compatibility
for use with SGE 5mL-100mL capacity syringe
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Cat. No. 509337 is a push-pull valve with removable 0.63 mm O.D. × 5 cm cone tip needle. This valve will fit removable needle syringes, 25μL-2.5mL.
Cat. No. 509345 is a push-pull with Kel-F male Luer lock termination to fit removable needle syringes, 25μL-2.5mL
Cat. No. 509353 is a push-button with Kel-F male Luer lock termination. This fits 5 mL to 2 liter syringes with removable Luer Lock or removable needle.
Cat. No. 509361 is a Luer Lock valve for any Luer Lock and Luer Tip syringe (50μl to 2 liter).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service