Z328405
BRAND® Transferpettor positive displacement pipette, digital volume
volume 200-1000 μL, accuracy: 0.5%, precision: 0.2%
About This Item
Recommended Products
feature
color-coded yellow
electronic: no
single-channel
DE-M marking
packaging
pack of 1 ea
manufacturer/tradename
BRAND 702806
volume measuring range
200-1000 μL, accuracy: 0.5%, precision: 0.2%
volume
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
- BRAND® Transferpettor digital volume pipette is utilized for the most demanding pipetting applications.
- Ideal for pipetting of highly viscous or foaming media, or media with high density or vapor pressure.
- The positive displacement principle enables the piston to be in direct contact with the aspirated liquid.
- The tightly fitting piston wipes the walls of the tips completely clean to provide accurate and reproducible results.
- No need to discard tips after each pipetting operation because of negligible residual wetting.
- The Transferpettor is suitable for media with density up to 13.6g/cm³, viscosity up to 140,000mm2/s, vapor pressure up to 500 mbar.
- Working temperature range: 15 °C to 40 °C.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service