WPB01040WA
Whirl-Pak® Thio-Bag®, Waste Water Sampling Bag
100 mL capacity
Synonym(s):
NELAC sample bag, sampling bag
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
sterility
sterile
packaging
pkg of 100 ea
manufacturer/tradename
Nasco B01040WA
parameter
-346-180 °F temp. range (-210 to 82 °C)
W × L × thickness
7.5 cm × 18.5 cm × 0.057 mm
capacity
100 mL
100 mL capacity
Related Categories
General description
Meets National Environmental Laboratory Accreditation Conference (NELAC) standard, June 2003.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Features and Benefits
- EPA approved
- Self-standing gusseted bag
- Sodium thiosulfate tablet in the bag for chlorine neutralization
- Built-in puncture proof closure
- Post-manufacturing sterilization for results you can trust.
Legal Information
Thio-Bag is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)