Skip to Content
Merck
All Photos(1)

Key Documents

UC180

Sigma-Aldrich

(S)-(+)-Nirvanol

Synonym(s):

(S)-(+)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, (S)-(+)-5-Ethyl-5-phenylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

off-white

solubility

DMF: soluble

storage temp.

2-8°C

SMILES string

CC[C@]1(NC(=O)NC1=O)c2ccccc2

InChI

1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)/t11-/m0/s1

InChI key

UDTWZFJEMMUFLC-NSHDSACASA-N

Biochem/physiol Actions

5-ethyl-5-phenylhydantoin (EPH) is a long-acting sedative. It is implicated to favor hepatocellular and thyroid follicular cell tumor progression.
CYP2B6 metabolite of (R)-(-)-mephenytoin; anticonvulsant; hypnotic.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Nirvanol is soluble in DMF.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pezeshk et al.
Journal of inorganic biochemistry, 42(4), 267-272 (1991-06-01)
The preparation and spectral properties of copper(II) complexes of two hydantoins are reported. Complexes of the general formula Cu(hyd)2(py)2, where hyd = phenytoin or nirvanol; and py = pyridine were prepared and characterized by infrared and ESR. Spectral data show
B F Bourgeois et al.
Epilepsia, 27(4), 412-418 (1986-07-01)
Stereoselective metabolism has been demonstrated for mephenytoin (MHT), R-MHT being demethylated to the pharmacologically active metabolite 5-phenyl-5-ethylhydantoin (PEH; nirvanol), and S-MHT undergoing aromatic hydroxylation to 4-OH-MHT, with formation of an intermediate arene oxide metabolite. PEH is responsible for the therapeutic
H Fujioka et al.
Journal of pharmacobio-dynamics, 9(3), 303-314 (1986-03-01)
Disposition of 1-benzenesulfonyl-5,5-diphenylhydantoin (II) having a potent anti-inflammatory activity was compared with that of 5,5-diphenylhydantoin (I), an antiepileptic drug, in order to elucidate whether the pharmacodynamic difference between them can be explained by their physicochemical and pharmacokinetic properties. After oral
H Heyn et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(9), 948-954 (1996-09-01)
In vitro methods were used to identify the cytochrome P450 (CYP) enzyme(s) involved in S-mephenytoin N-demethylation. S-Mephenytoin (200 microM) was incubated with human liver microsomes, and nirvanol formation was quantitated by reversed-phase HPLC. S-Mephenytoin N-demethylase activity in a panel of
R W Nims et al.
Journal of biochemical toxicology, 9(6), 279-288 (1994-12-01)
To explore the enantioselectivity of ligand interaction with the putative phenobarbital receptor, the pharmacodynamics of cytochrome P450 2B (CYP2B) induction by racemic 5-ethyl-5-phenylhydantoin and its two enantiomers were investigated in the male F344/NCr rat and in cultured adult male rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service