Skip to Content
Merck
All Photos(1)

Key Documents

U60100-HH

Sigma-Aldrich

DMT-rU-CPG

pore size 500 Å, 25-35 μmol/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

product line

Proligo Reagents

Assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

packaging

pkg of 1 × 1 g U601001-01

pore size

500 Å

capacity

≥95.0 % (Coupling Efficiency)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service