SRP8046
Netrin-1 (human): FC (human)
recombinant, expressed in HEK 293 cells, >95% (SDS-PAGE)
Synonym(s):
ACRP30headless:TNFSF13, NTN1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
Assay
>95% (SDS-PAGE)
form
lyophilized
mol wt
~90 kDa by SDS-PAGE
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
impurities
<0.1 EU/μg endotoxin, tested
color
white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... NTN1(9423)
General description
Netrin-1 controls guidance of CNS commissural axons and peripheral motor axons. Its association with either DCC or some UNC5 receptors will lead to axon attraction or repulsion, respectively. It also serve as a survival factor via its association with its receptors which prevent the initiation of apoptosis. Netrin 1 is also Involved in tumorigenesis by regulating apoptosis.
Physical form
Lyophilized from PBS with trehalose and Brij35.
Reconstitution
Reconstitute with 100 μL deionized water.
Other Notes
Human netrin-1 (aa 25-604) is fused at the C-terminus to the Fc portion of human IgG1.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service