Skip to Content
Merck
All Photos(1)

Key Documents

SRP3025

Sigma-Aldrich

sDLL-1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

Delta-1, Delta-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

3.0-5.0 μg/mL

mol wt

57.0-60.0 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DLL1(28514)

General description

Human sDLL-1 comprises the extracellular signaling domain of DLL-1, a member of a structurally-related family of single-pass type I trans-membrane proteins that serve as ligands for Notch receptors. It is expressed in the heart and pancreas, and to a lesser extent in various other tissues. DLL-1 functions to specifically activate the Notch-1 and Notch-2 receptors. The Notch signaling pathway regulates endothelial-cell differentiation, proliferation and apoptosis, and is essential for the development, maintenance and remodeling of the vascular system. DLL-1 suppresses differentiation of hematopoietic progenitor cells into the B-cell lineage while promoting differentiation to T-cell and NK cell precursors. Recombinant human sDLL-1 is a 57.0-60.0 kDa glycoprotein containing 522 amino-acid residues

Biochem/physiol Actions

Human sDLL-1 comprises the extracellular signaling domain of DLL-1, a member of a structurally-related family of single-pass type I trans-membrane proteins that serve as ligands for Notch receptors. It is expressed in the heart and pancreas, and to a lesser extent in various other tissues. Recombinant human sDLL-1 is a 57.0-60.0 kDa glycoprotein containing 522 amino-acid residues

Sequence

SGVFELKLQE FVNKKGLLGN RNCCRGGAGP PPCACRTFFR VCLKHYQASV SPEPPCTYGS AVTPVLGVDS FSLPDGGGAD SAFSNPIRFP FGFTWPGTFS LIIEALHTDS PDDLATENPE RLISRLATQR HLTVGEEWSQ DLHSSGRTDL KYSYRFVCDE HYYGEGCSVF CRPRDDAFGH FTCGERGEKV CNPGWKGPYC TEPICLPGCD EQHGFCDKPG ECKCRVGWQG RYCDECIRYP GCLHGTCQQP WQCNCQEGWG GLFCNQDLNY CTHHKPCKNG ATCTNTGQGS YTCSCRPGYT GATCELGIDE CDPSPCKNGG SCTDLENSYS CTCPPGFYGK ICELSAMTCA DGPCFNGGRC SDSPDGGYSC RCPVGYSGFN CEKKIDYCSS SPCSNGAKCV DLGDAYLCRC QAGFSGRHCD DNVDDCASSP CANGGTCRDG VNDFSCTCPP GYTGRNCSAP VSRCEHAPCH NGATCHERGH RYVCECARGY GGPNCQFLLP ELPPGPAVVD LTEKLEGQGG PF

Physical form

Lyophilized from 1x PBS, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin G Leong et al.
Differentiation; research in biological diversity, 76(6), 699-716 (2008-06-21)
The Notch family of transmembrane receptors are important mediators of cell fate determination. Accordingly, Notch signaling is intimately involved in the development of numerous tissues. Recent findings have highlighted a critical role for Notch signaling in normal prostate development. Notch
Renée F de Pooter et al.
Methods in molecular biology (Clifton, N.J.), 380, 73-81 (2007-09-19)
Mature hematopoietic cells, like all other terminally differentiated lineages, arise during ontogeny via a series of increasingly restricted intermediates. Hematopoietic progenitors derive from the mesoderm, which gives rise to hemangioblasts that can differentiate into endothelial or endocardial precursors, or hematopoietic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service