Skip to Content
Merck
All Photos(1)

Key Documents

SML3657

Sigma-Aldrich

uPSEM817 tartrate

≥98% (HPLC)

Synonym(s):

2-N-Propoxy-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline tartrate, PSEM 817 tartrate, Ultrapotent pharmacologically selective effector molecule 817 tararate, uPSEM817 tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O·C4H6O6
CAS Number:
Molecular Weight:
419.43
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ultrapotent PSEM agonist for chimeric ligand-gated ion channel pores with nAChR α7 LBD L181G/Q139L/Y217F mutant (PSAM4) in vitro and in vivo.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrapotent chemogenetics for research and potential clinical applications
Science (New York, N.Y.), 364(6436) (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service