Skip to Content
Merck
All Photos(1)

Key Documents

SML3642

Sigma-Aldrich

PD 173955

≥98% (HPLC)

Synonym(s):

6-(2,6-Dichlorophenyl)-8-methyl-2-[[3-(methylthio)phenyl]amino]pyrido[2,3-d]pyrimidin-7(8H)-one, PD-173955

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16Cl2N4OS
CAS Number:
Molecular Weight:
443.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

SMILES string

S(C)c1cc(ccc1)Nc2nc3c(cn2)C=C(C(=O)N3C)c4c(cccc4Cl)Cl

InChI

1S/C21H16Cl2N4OS/c1-27-19-12(9-15(20(27)28)18-16(22)7-4-8-17(18)23)11-24-21(26-19)25-13-5-3-6-14(10-13)29-2/h3-11H,1-2H3,(H,24,25,26)

InChI key

VAARYSWULJUGST-UHFFFAOYSA-N

Biochem/physiol Actions

PD 173955 is Tyrosine kinase inhibitor (TKI), which potently inhibits Bcr-Abl (IC50 = 1-2 nM), Src family kinase inhibitor specific for Src and Yes kinases (IC50 of 22 nM), and MAPK6/ERK3. According to IUPHAR/BPS Guide to PHARMACOLOGY PD 173955 is a best standard MAPK6 inhibitor pKD = 6.8
Tyrosine kinase type I inhibitor; potently inhibits Bcr-Abl and MAPK6/ERK3; Src family kinase inhibitor specific for Src and Yes kinases

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arjun Kumar Kalimuthu et al.
Scientific reports, 11(1), 21488-21488 (2021-11-04)
Spice-rich recipes are referred to as "functional foods" because they include a variety of bioactive chemicals that have health-promoting properties, in addition to their nutritional value. Using pharmacoinformatics-based analysis, we explored the relevance of bioactive chemicals found in Rasam (a
Anjana Sinha et al.
ACS medicinal chemistry letters, 10(10), 1430-1435 (2019-10-18)
Compound 3a, DV2-103, is a kinase inactive analogue of a potent Abl1/Src kinase inhibitor, PD173955, 2. Both compounds, 2 and 3a, are known to reduce production of beta amyloid (Aβ) peptide in cells and animal models. We have now prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service