Skip to Content
Merck
All Photos(1)

Key Documents

SML3579

Sigma-Aldrich

Pyrimethamine

≥98% (HPLC)

Synonym(s):

2,4-Diamino-5-(4-chlorophenyl)-6-ethylpyrimidine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13ClN4
CAS Number:
Molecular Weight:
248.71
MDL number:
UNSPSC Code:
12352116
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Wramed)

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)c2c(nc(nc2CC)N)N

InChI

1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)

InChI key

WKSAUQYGYAYLPV-UHFFFAOYSA-N

Biochem/physiol Actions

Antiparasitic inhibitor of dihydrofolate reductase; folic acid antagonist
Pyrimethamine is an antiparasitic drug used to treat malaria, toxoplasmosis and cystoisosporiasis. Pyrimethamine is a potent inhibitor of dihydrofolate reductase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The multifaceted antineoplastic role of pyrimethamine against human malignancies
Shanaya Ramchandani
IUBMB Life, 74, 198-212 (2022)
Mutations in Plasmodium falciparum dihydrofolate reductase and dihydropteroate synthase and epidemiologic patterns of pyrimethamine-sulfadoxine use and resistance
C V Plowe
The Journal of Infectious Diseases, 176, 1590-1596 (1997)
Lisa N Heppler et al.
The Journal of biological chemistry, 298(2), 101531-101531 (2021-12-27)
Cancer is often characterized by aberrant gene expression patterns caused by the inappropriate activation of transcription factors. Signal transducer and activator of transcription 3 (STAT3) is a key transcriptional regulator of many protumorigenic processes and is persistently activated in many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service