Skip to Content
Merck
All Photos(1)

Key Documents

SML3086

Sigma-Aldrich

Diclofenac

≥98% (HPLC), powder, COX inhibitor

Synonym(s):

2-(2,6-dichloranilino) phenylacetic acid, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11Cl2NO2
CAS Number:
Molecular Weight:
296.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Diclofenac, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(Cl)=C1NC2=CC=CC=C2CC(O)=O

InChI

1S/C14H11Cl2NO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17H,8H2,(H,18,19)

InChI key

DCOPUUMXTXDBNB-UHFFFAOYSA-N

Biochem/physiol Actions

Diclofenac is an orally available, potent and selective nonsteroidal anti-inflammatory drug (NSAID). Diclofenac is used to treat pain and inflammatory diseases. It inhibits both cycloxygenase-1 (COX-1) and cycloxygenase-2 (COX-2).
anti-inflammatory, antipyretic, analgesic, COX inhibitor, NSAID

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Yoshida et al.
Bioorganic & medicinal chemistry, 20(19), 5705-5719 (2012-09-11)
Dipeptidyl peptidase IV (DPP-4) inhibition is suitable mechanism for once daily oral dosing regimen because of its low risk of hypoglycemia. We explored linked bicyclic heteroarylpiperazines substituted at the γ-position of the proline structure in the course of the investigation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service