Skip to Content
Merck
All Photos(1)

Key Documents

SML2762

Sigma-Aldrich

RG1534

≥98% (HPLC)

Synonym(s):

2-Amino-N-((4-((5-chloropyrimidin-2-yl)oxy)-3-methylphenyl)carbamoyl)benzamide, 2-Amino-N-[[[4-[(5-chloro-2-pyrimidinyl)oxy]-3-methylphenyl]amino]carbonyl]benzamide;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClN5O3
CAS Number:
Molecular Weight:
397.82
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=CC=CC=C1C(NC(NC2=CC(C)=C(C=C2)OC3=NC=C(C=N3)Cl)=O)=O

InChI

1S/C19H16ClN5O3/c1-11-8-13(6-7-16(11)28-19-22-9-12(20)10-23-19)24-18(27)25-17(26)14-4-2-3-5-15(14)21/h2-10H,21H2,1H3,(H2,24,25,26,27)

InChI key

NLVSCQFSPVSAOF-UHFFFAOYSA-N

Biochem/physiol Actions

RG1534 is an inhibitor of hTERT promoter activity that selectively targets G-hairpin stem loop of the human telomerase reverse transcriptase gene (hTERT) that downregulates hTERT expression. RG1534 induces mitochondrial defects, oxidative stress, DNA damage and apoptosis in cancer cell while sparing normal cells
inhibitor of hTERT promoter activity that selectively targets G-hairpin stem loop of the human telomerase reverse transcriptase gene (hTERT)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin H Song et al.
Cell chemical biology, 26(8), 1110-1121 (2019-06-04)
Increased telomerase activity is associated with malignancy and poor prognosis in human cancer, but the development of targeted agents has not yet provided clinical benefit. Here we report that, instead of targeting the telomerase enzyme directly, small molecules that bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service