Skip to Content
Merck
All Photos(1)

Key Documents

SML2657

Sigma-Aldrich

MP-A08

≥98% (HPLC)

Synonym(s):

4-Methyl-N-[2-[[[2-[[(4-methylphenyl)sulfonyl]amino]phenyl]imino]methyl]phenyl]benzenesulfonamide, NSC-122314

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25N3O4S2
CAS Number:
Molecular Weight:
519.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2c(cccc2)\N=C\c3c(cccc3)N[S](=O)(=O)c4ccc(cc4)C)c1ccc(cc1)C

InChI key

FLDBNMYJUMAXDQ-TURZUDJPSA-N

Biochem/physiol Actions

MP-A08 is a highly selective ATP-competitive sphingosine kinase inhibitor that targets both SK1 and SK2. It is more selective and acts by different mechanisms than other SK inhibitors. MP-A0 targets the ATP-binding pocket, rather than the sphingosine binding pocket, with Ki values of 6.9 and 27 μM, respectively. Also, both SKI-II and PF-543 are thought to act in part by targeting sphingosine kinase for proteasomal degradation, which is not the case with MP-A08. MP-A08 was found to suppress the growth of human lung tumor xenografts in mice, both inducing apoptosis and inhibiting angiogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service