Skip to Content
Merck
All Photos(1)

Key Documents

SML2255

Sigma-Aldrich

BI-2545

≥98% (HPLC)

Synonym(s):

(1R,5S,6S)-3,5-bis(Trifluoromethyl)benzyl 6-((1H-benzo[d][1,2,3]triazole-5-carboxamido)methyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate, [3,5-bis(Trifluoromethyl)phenyl]methyl (1R,5S,6S)-6-{[(1H-1,2,3-benzotriazol-5-yl)formamido]methyl}-3-azabicyclo[3.1.0]hexane-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19F6N5O3
CAS Number:
Molecular Weight:
527.42
UNSPSC Code:
41106609
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1C[C@]2([H])[C@@H](CNC(C3=CC=C(NN=N4)C4=C3)=O)[C@]2([H])C1)OCC5=CC(C(F)(F)F)=CC(C(F)(F)F)=C5

InChI

1S/C23H19F6N5O3/c24-22(25,26)13-3-11(4-14(6-13)23(27,28)29)10-37-21(36)34-8-16-15(17(16)9-34)7-30-20(35)12-1-2-18-19(5-12)32-33-31-18/h1-6,15-17H,7-10H2,(H,30,35)(H,31,32,33)/t15-,16-,17+

InChI key

ZDOBSAPHUUUOHX-OSYLJGHBSA-N

Biochem/physiol Actions

Autotaxin (ATX; ENPP2) inhibitor with improved potency, oral availability, and lysophosphatidic acid (LPA)-lowering efficacy in rats than PF-8380.
BI-2545 is a PF-8380 structure analog with improved autotaxin (ATX; ENPP2) inhibitory potency (human/rat ATX IC50 = 2.2/3.4 nM, IC50 = 29/96 nM using human/rat whole blood (WB); human ATX/rat ATX/rat WB IC50 = 6.5/13/307 nM with PF-8380), in vivo pharmacokinetic profile, oral availability, and lysophosphatidic acid (LPA)-lowering efficacy (BI-2545 Cmax/AUC/T1/2/Max 18:2 LPA reduction = 140 nM/675 nM•h/3.4 h/90.4% post 10 mg/kg p.o. in rats vs. 60.3 nM/342 nM•h/2.5 h/32.2% with PF-8380). BI-2545 exhibits no hERG inhibitory activity (IC50 >10 μM vs. 480 nM with PF-8380) and displays significant cross-reactivity towards only 4 targets among a panel of 48 enzymes/receptors/transporters/channel proteins at a high concentration of 10 μM (55%, 80%, 66%, 61% inhibtion of 5-HT2a, L-type Calcium channel, Na+ channel site 2, and norepinephrine transporter, respectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian A Kuttruff et al.
ACS medicinal chemistry letters, 8(12), 1252-1257 (2017-12-21)
In an effort to find new therapeutic interventions addressing the unmet medical need of patients with idiopathic pulmonary fibrosis, we initiated a program to identify new autotaxin (ATX) inhibitors. Starting from a recently published compound (PF-8380), we identified several highly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service