Skip to Content
Merck
All Photos(1)

Key Documents

SML2219

Sigma-Aldrich

SGC-AAK1-1

≥98% (HPLC)

Synonym(s):

N-(6-(3-(N,N-Diethylsulfamoylamino)phenyl)-1H-indazol-3-yl)cyclopropanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N5O3S
CAS Number:
Molecular Weight:
427.52
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)S(NC1=CC=CC(C2=CC(NN=C3NC(C4CC4)=O)=C3C=C2)=C1)(=O)=O

Biochem/physiol Actions

SGC-AAK1-1 is an ATP-competitive kinase inhibitor against AP2-associated protein kinase 1/AAK1 and BMP-2-inducible protein kinase/BIKE/BMP2K (Ki = 9.1 & 17 nM, respectively, by ATP site fluorescent tracer displacement assay; AAK1 IC50 = 270 nM by coupled enzyme assay). SGC-AAK1-1 downregulates cellular AP2M1 Thr156 phosphorylation level in a dose-dependent manner (ECmax ∼12.5 μM) without detectable cytotoxicity. Selectivity profiling identifies only 5 additional kinases among a panel of ∼400 with >50% inhibition by 1 μM SGC-AAK1-1 (KD = 72 nM/RIOK1, 290 nM/RIOK3, 260 nM/PIP5K1C, 880 nM/CDKL1, 960 nM/MYLK2 by DiscoverX KINOMEscan). SGC-AAK1-1N is a structure analog and the recommended negative control (IC50 = 11 μM by AAK1 activity assay; KD = 8.8 μM/AAK1 & >10 μM/BIKE vs. KD = 26 nM/AAK1 & 930 nM/BIKE with SGC-AAK1-1 by DiscoverX). For characterization details of SGC-AAK1-1, please visit the SGC-AAK1-1 probe summary on the Structural Genomics Consortium (SGC) website.

SGC-AAK1-1N is the negative control for the active probe, SGC-AAK1-1. SGC-AAK1-1N is available from Sigma. To learn more about and purchase SGC-AAK1-1N, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

SGC-AAK1-1 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service