Skip to Content
Merck
All Photos(1)

Key Documents

SML2054

Sigma-Aldrich

SEA0400

≥98% (HPLC)

Synonym(s):

SEA0400, 2-[4-[(2,5-Difluorophenyl)methoxy]phenoxy]-5-ethoxybenzenamine, SEA 0400

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19F2NO3
CAS Number:
Molecular Weight:
371.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

shipped in

ambient

storage temp.

−20°C

SMILES string

CCOC1=CC=C(OC2=CC=C(OCC3=C(F)C=CC(F)=C3)C=C2)C(N)=C1

InChI

1S/C21H19F2NO3/c1-2-25-18-8-10-21(20(24)12-18)27-17-6-4-16(5-7-17)26-13-14-11-15(22)3-9-19(14)23/h3-12H,2,13,24H2,1H3

InChI key

YSUBLPUJDOWYDP-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective inhibitor of Na+−Ca++ exchanger (NCX)
SEA0400 is a potent and selective inhibitor of Na+/Ca2+exchanger (NCX). SEA0400 potently inhibits Na+/Ca2+ exchanger activity in rat astrocytes, microglia, cultured neurons, and rat cardiomyocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Matsuda et al.
The Journal of pharmacology and experimental therapeutics, 298(1), 249-256 (2001-06-16)
The effect of the newly synthesized compound 2-[4-[(2,5-difluorophenyl)methoxy]phenoxy]-5-ethoxyaniline (SEA0400) on the Na+-Ca2+ exchanger (NCX) was investigated and compared against that of 2-[2-[4-(4-nitrobenzyloxy)phenyl]ethyl]isothiourea (KB-R7943). In addition, the effects of SEA0400 on reperfusion injury in vitro and in vivo were examined. SEA0400
Kanna Yamashita et al.
Naunyn-Schmiedeberg's archives of pharmacology, 389(11), 1205-1214 (2016-08-03)
Recently, YM-244769 (N-(3-aminobenzyl)-6-{4-[(3-fluorobenzyl)oxy]phenoxy} nicotinamide) has been reported as a new potent and selective Na+/Ca2+ exchange (NCX) inhibitor by using various cells transfected with NCX using the 45Ca2+ fluorescent technique. However, the electrophysiological study of YM-244769 on NCX had not been
Torsten Christ et al.
European journal of pharmacology, 788, 286-293 (2016-07-05)
The Na(+)/Ca(2+) exchanger (NCX) plays a major role in myocardial Ca(2+) homoeostasis, but is also considered to contribute to the electrical instability and contractile dysfunction in chronic atrial fibrillation (AF). Here we have investigated the effects of the selective NCX

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service