Skip to Content
Merck
All Photos(1)

Key Documents

SML1976

Sigma-Aldrich

DIM-C-pPhCO2Me

≥98% (HPLC)

Synonym(s):

4-(di-1H-Indol-3-ylmethyl)-benzoic acid methyl ester, Benzoic acid, 4-(di-1H-indol-3-ylmethyl)-, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N2O2
CAS Number:
Molecular Weight:
380.44
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

pink to light red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC)C1=CC=C(C(C2=CNC3=C2C=CC=C3)C4=CNC5=CC=CC=C54)C=C1

InChI

1S/C25H20N2O2/c1-29-25(28)17-12-10-16(11-13-17)24(20-14-26-22-8-4-2-6-18(20)22)21-15-27-23-9-5-3-7-19(21)23/h2-15,24,26-27H,1H3

InChI key

BBAOKSZCULLDIW-UHFFFAOYSA-N

Biochem/physiol Actions

DIM-C-pPhCO2Me is a NR4A1 (Nur77) antagonist that exhibit potent anticancer activity in pancreatic, colon, breast, kidney, and rhabdomyosarcoma cells. DIM-C-pPhCO2Me decreases expression of PAX3-FOXO1A and β1-integrin proteins in Rh30 rhabdomyosarcoma and MDA-MB-231 breast cancer cells. DIM-C-pPhCO2Me inhibits migration of Rh30 and MDA-MB-231 cancer cells.
DIM-C-pPhCO2Me is also known as 1,1-bis(3′-indolyl)-1-(p-carboxymethylphenyl)methane. In pancreatic and colon cancer cells, DIM-C-pPhCO2Me plays an inhibitory role on cell migration. DIM-C-pPhCO2Me prevents the action of mechanistic target of rapamycin (mTOR) and reduces the multiplication and survival of cancer cells. It stimulates reactive oxygen species (ROS) in various cancer cell lines.
NR4A1 (Nur77) antagonist that exhibit potent anticancer activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Hedrick et al.
Molecular and cellular biology, 36(9), 1383-1394 (2016-03-02)
Overexpression of the nuclear receptor 4A1 (NR4A1) in breast cancer patients is a prognostic factor for decreased survival and increased metastasis, and this has been linked to NR4A1-dependent regulation of transforming growth factor β (TGF-β) signaling. Results of RNA interference
The nuclear orphan receptor Nr4a1 regulates beta1-integrin expression in pancreatic and colon cancer cells and can be targeted by Nr4a1 antagonists
Hedrick E, et al.
Molecular Carcinogenesis, 56(9), 2066-2075 (2017)
Alexandra Lacey et al.
Cancer research, 77(3), 732-741 (2016-11-20)
Alveolar rhabdomyosarcoma (ARMS) is a devastating pediatric disease driven by expression of the oncogenic fusion gene PAX3-FOXO1A. In this study, we report overexpression of the nuclear receptor NR4A1 in rhabdomyosarcomas that is sufficient to drive high expression of PAX3-FOXO1A there.
Syng-Ook Lee et al.
Molecular endocrinology (Baltimore, Md.), 28(10), 1729-1739 (2014-08-08)
1,1-Bis(3'-indolyl)-1-(p-substituted phenyl)methane (C-DIM) compounds exhibit antineoplastic activity in multiple cancer cell lines and the p-hydroxyphenyl analog (DIM-C-pPhOH) inactivates nuclear receptor 4A1 (NR4A1) in lung and pancreatic cancer cell lines. Using a series of 14 different p-substituted phenyl C-DIMs, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service