Skip to Content
Merck
All Photos(1)

Key Documents

SML1737

Sigma-Aldrich

Beraprost sodium

≥98% (HPLC)

Synonym(s):

2,3,3a,8b-Tetrahydro-2-hydroxy-1-(3-hydroxy-4-methyl-1-octen-6-ynyl)-1H-cyclopenta(b)benzofuran-5-butanoic acid sodium salt, Procyclin, TRK 100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29O5 · Na
CAS Number:
Molecular Weight:
420.47
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[H][C@]12[C@](C[C@H](O)[C@H]2/C=C/[C@H](O)C(CC#CC)C)([H])OC3=C1C=CC=C3CCCC([O-])=O.[Na+]

InChI

1S/C24H30O5.Na/c1-3-4-7-15(2)19(25)13-12-17-20(26)14-21-23(17)18-10-5-8-16(24(18)29-21)9-6-11-22(27)28;/h5,8,10,12-13,15,17,19-21,23,25-26H,6-7,9,11,14H2,1-2H3,(H,27,28);/q;+1/p-1/b13-12+;

InChI key

YTCZZXIRLARSET-UEIGIMKUSA-M

General description

Beraprost sodium is an orally active prostacyclin analog.

Biochem/physiol Actions

Beraprost sodium is a chemically stable analog of prostacyclin PGI2.
Beraprost sodium is a chemically stable analog of prostacyclin PGI2. As other prostanoids, beraprost is a potent vasodilator and possesses antithrombotic, antiproliferative and anti-inflammatory properties. Beraprost sodium activates prostaglandin I (IP) receptor, which is coupled with Gs proteins and activates adenylate cyclase. It has been studied for the treatment of pulmonary hypertension.
Beraprost sodium is used as a therapeutic agent for chronic artery obstructions or primary pulmonary hypertension. In individuals with coronary artery disease, beraprost sodium helps to reduce oxidative stress and developed forearm endothelium dependent vasodilation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin Resistance: New Insights for the Healthcare Professional (2013)
Textbook of Critical Care (2011)
Aging and Age-Related Disorders (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service