Skip to Content
Merck
All Photos(1)

Key Documents

SMB01020

Sigma-Aldrich

26-Desglucoavenacoside A

≥85% (LC/MS-ELSD)

Synonym(s):

26-Deglucoavenacoside A, Deglucoavenacoside A, Desglucoavenacoside A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H72O18
CAS Number:
Molecular Weight:
901.04
UNSPSC Code:
12352205
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

901.04

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O[C@@H]9O[C@H]([C@@H]([C@H]([C@H]9O)O)O)C)CO)O[C@H]3CC[C@@]4(C5[C@H](C6[C@@]([C@@H]7[C@@H](O[C@@]8(O[C@@](CC8)(CO)C)[C@H]7C)C6)(CC5)C)CC=C4C3)C

InChI key

XUUBSPXZCBAWET-AACVHOIVSA-N

General description

26-desglucoavenacoside A is a steroidal saponin commonly available from Avena sativa (oats). Current research indicates that this naturally occurring metabolite may possesses diverse biological activities, including anti-parasitic and antimicrobial properties.

Application

Desglucoavenacoside A is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service