Skip to Content
Merck
All Photos(2)

Documents

SAB5200104

Sigma-Aldrich

Anti-SLC12A3 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-NCC, Anti-Slc12a3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 160 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 12 member 3 (SLC12A3) is also known as thiazide-sensitive Na+-Cl- cotransporter (NCCT). It is expressed in the apical cells of the distal convoluted tubule. The gene encoding this 1021 amino acid protein is localized on human chromosome 16q13. The gene ID for the protein is 6559.

Specificity

Detects ~160 kDa.

Immunogen

AA74-95 (rat), 76-97 (hum)

Biochem/physiol Actions

Solute carrier family 12 member 3 (SLC12A3) is one of the major components of the sodium chloride reabsorption pathway. It controls the Cl- concentration and is also involved in the transepithelial ion absorption and secretion. Mutations in the gene encoding SLC12A3 have been associated with Gitelman syndrome.
This Na-Cl cotransporter is associated with maintaining blood pressure.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variants of SLC12A3 modulate serum lipid profiles in a group of Mongolian pedigree population
An C, et al.
Lipids in Health and Disease, 17(1), 83-83 (2018)
Linghong Wang et al.
Acta biochimica et biophysica Sinica, 47(5), 325-334 (2015-04-07)
The thiazide-sensitive Na(+)-Cl(-) cotransporter (TSC) is responsible for the major sodium chloride reabsorption pathway, which is located in the apical membrane of the epithelial cells of the distal convoluted tubule. TSC is involved in several physiological activities including transepithelial ion
Jiewei Luo et al.
Endocrine journal, 62(1), 29-36 (2014-10-03)
Gitelman syndrome (GS) is a salt-wasting tubulointerstitial disease of autosomal recessive inheritance (OMIM613395) caused by genic mutation of SLC12A3, which codes thiazide-sensitive Na-Cl cotransporter (NCCT) gene. The gene mutation of the majority of GS patients is compound heterozygous. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service