Skip to Content
Merck
All Photos(7)

Key Documents

SAB4503824

Sigma-Aldrich

Anti-phospho-p130 Cas (pTyr410) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 93 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr410)

Gene Information

human ... BCAR1(9564)

General description

Breast cancer antiestrogen resistance 1 (BCAR1), also known as p130 Cas, is encoded by the gene mapped to human chromosome 16q23. BCAR1 belongs to Cas (Crk-associated substrate) protein family. The protein is characterized with an amino-terminal Src homology 3 (SH3) domain, a Src-binding domain, a large substrate domain, and a helix-loop-helix domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human p130 Cas around the phosphorylation site of Tyr410.

Immunogen Range: 376-425

Biochem/physiol Actions

Breast cancer antiestrogen resistance 1 (BCAR1), also known as p130 Cas, plays a vital role in various cellular processes such as migration, adhesion, cytoskeleton reorganization host-pathogen interaction, survival, proliferation, transformation, and possibly metastasis. It is associated with the growth factor receptor EGFR and the metastasis suppressor KAI1, in vitro. Overexpression of BCAR1 in localized prostate cancers (LPC) suggests that this protein has potential as a marker for both prognosis and treatment. It is also considered to be a potential marker for breast cancer. Neural precursor cell expressed developmentally down-regulated protein 9 (NEDD-9) and BCAR1, via SRC, promote removal of E-Cadherin from cell membrane, and trigger E-cadherin degradation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaëlle Fromont et al.
The Prostate, 67(3), 268-273 (2006-12-29)
The 16q23 locus has been recently suggested in both breast and prostate cancer to contain a gene involved in disease progression. The breast cancer antiestrogen resistance 1 (BCAR1) gene, located at 16q23, contributes to many cellular processes including migration and
Lambert C J Dorssers et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 10(18 Pt 1), 6194-6202 (2004-09-28)
BCAR1, the human homologue of the rat p130Cas protein, was identified in a functional screen for human breast cancer cell proliferation resistant to antiestrogen drugs. Here, we study the prognostic value of quantitative BCAR1 levels in a large series of
Nadezhda Tikhmyanova et al.
PloS one, 6(7), e22102-e22102 (2011-07-19)
The Cas scaffolding proteins (NEDD9/HEF1/CAS-L, BCAR1/p130Cas, EFSSIN, and HEPL/CASS4) regulate cell migration, division and survival, and are often deregulated in cancer. High BCAR1 expression is linked to poor prognosis in breast cancer patients, while upregulation of NEDD9 contributes to the
Bo Deng et al.
Annals of surgical oncology, 20 Suppl 3, S701-S708 (2013-08-02)
This study was designed to determine the prognostic value of BCAR1 expression and its associations with clinical-demographical characteristics in multiple centers of non-small cell lung cancer (NSCLC) patients. Gene expression microarray (mRNA) of 77 adenocarcinomas from Mayo Clinic, RNA-sequencing of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service