Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500230

Sigma-Aldrich

Anti-SARM antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-SARM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 80 kDa

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SARM1(23098)

General description

The gene SARM1 (sterile α and TIR motif containing 1) is mapped to human chromosome 17q11. It is mainly expressed in neurons. The encoded protein belongs to the MyD88 (myeloid differentiation primary response gene 88) family of adaptor proteins. The protein contains a armadillo repeat region, SAM (sterile α motif) domains and a TIR (toll-interleukin receptor) domain. It localizes in the mitochondria as well as nucleus.

Immunogen

SARM antibody was raised against a peptide corresponding to amino acids near the C-terminus of human SARM.

Biochem/physiol Actions

SARM1 (sterile α and TIR motif containing 1) is a TIR (toll-interleukin receptor) adaptor protein. It plays a crucial role in the immune system and TLR (toll like receptor) signaling. It interacts with PINK1 (PTEN-induced putative kinase 1) and is involved in the stabilization of PINK1 on depolarized mitochondria. During inflammation-mediated apoptosis, the protein is involved in the stabilization of nuclear lamins. SARM1 is also associated with Wallerian degeneration.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500230.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Zhou et al.
Developmental and comparative immunology, 39(1-2), 117-126 (2012-03-01)
Toll-like receptors (TLRs) are important pattern-recognition receptors (PRRs) that trigger innate immune response and mediate acquired immunity. Evidence has shown that SARM1 (sterile-α and TIR motif containing protein 1) is one of five TIR domain-containing adaptor proteins involved in TLRs
Laura Conforti et al.
Nature reviews. Neuroscience, 15(6), 394-409 (2014-05-21)
Axon degeneration is a prominent early feature of most neurodegenerative disorders and can also be induced directly by nerve injury in a process known as Wallerian degeneration. The discovery of genetic mutations that delay Wallerian degeneration has provided insight into
Chad R Sethman et al.
PloS one, 8(7), e70994-e70994 (2013-08-08)
Sterile alpha and armadillo-motif containing protein (SARM), a highly conserved and structurally unique member of the MyD88 family of Toll-like receptor adaptors, plays an important role in innate immunity signaling and apoptosis. Its exact mechanism of intracellular action remains unclear.
Hitoshi Murata et al.
Molecular biology of the cell, 24(18), 2772-2784 (2013-07-26)
Mutations in PTEN-induced putative kinase 1 (PINK1) or parkin cause autosomal recessive forms of Parkinson's disease. Recent work suggests that loss of mitochondrial membrane potential stabilizes PINK1 and that accumulated PINK1 recruits parkin from the cytoplasm to mitochondria for elimination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service