Skip to Content
Merck
All Photos(3)

Key Documents

SAB2702160

Sigma-Aldrich

Monoclonal Anti-RPA2 antibody produced in mouse

Synonym(s):

REPA2, RPA32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12F3.3
monoclonal

form

liquid

mol wt

32 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA2(6118)

General description

Replication protein A2 (RPA2) is a 32kDa protein. The gene encoding it is localized on human chromosome 1p35.

Immunogen

Full-length human RPA-32 expressed in E. coli.

Application

For ICC/IF: Use at a dilution of 1:100-1:1000. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500-1:3000. Predicted molecular weight: 29 kDa. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

Replication protein A2 (RPA2) is involved in DNA repair and replication. The protein has been shown to be phosphorylated as a response to DNA damage and apoptosis in vitro.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, pH 7.4, containing no preservatives

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic analysis of BRCA1-depleted cell line reveals a putative role for replication protein A2 up-regulation in BRCA1 breast tumor development.
Bouley J
Proteomics. Clinical Applications, 4(5), 489-498 (2010)
Loss of heterozygosity at chromosome 1p in different solid human tumours: association with survival.
Ragnarsson G
British Journal of Cancer, 79(9-10), 1468-1474 (1999)
Phosphorylation of replication protein A middle subunit (RPA32) leads to a disassembly of the RPA heterotrimer.
Treuner K
The Journal of Biological Chemistry, 274(22), 15556-15561 (1999)
Holger Stephan et al.
Nucleic acids research, 37(18), 6028-6041 (2009-08-13)
The human single-stranded DNA-binding protein, replication protein A (RPA), is regulated by the N-terminal phosphorylation of its 32-kDa subunit, RPA2. RPA2 is hyperphosphorylated in response to various DNA-damaging agents and also phosphorylated in a cell-cycle-dependent manner during S- and M-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service