Skip to Content
Merck
All Photos(2)

Documents

SAB1300928

Sigma-Aldrich

Anti-MARS2 (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-Methionine--tRNA ligase 2, Anti-Methionyl-tRNA synthetase, mitochondrial [Precursor], Anti-Mitochondrial methionine--tRNA ligase, Anti-MtMetRS, EC 6.1.1.10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARS2(92935)

General description

Methionine-tRNA ligase (EC 6.1.1.10) catalyzes the chemical reaction: ATP + L-methionine + tRNAMet ⇔ AMP + diphosphate + L-methionyl-tRNAMet The 3 substrates of this enzyme are ATP, L-methionine, and tRNA(Met), whereas its 3 products are AMP, diphosphate, and L-methionyl-tRNA(Met). This enzyme participates in 3 metabolic pathways: methionine metabolism, selenoamino acid metabolism, and aminoacyl-trna biosynthesis.

Immunogen

MARS2 (NP_612404, 568-603)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human MARS2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service