S6143
Sigma Pasteur pipettes
cotton plugged, L 9 in.
Synonym(s):
63B6009-00P, Pasteur Pipette, Pasteur Pipettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.01
Recommended Products
material
glass
sterility
sterile; γ-irradiated
sterile; γ-irradiated
feature
graduations: no
plugged
packaging
pack of 25 ea (acked in blisterpacks)
manufacturer/tradename
Sigma-Aldrich
L
9 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service