Skip to Content
Merck
All Photos(4)

Key Documents

PLN350

Sigma-Aldrich

GenElute Plasmid Miniprep Kit

greener alternative

sufficient for 350 purifications

Synonym(s):

GenElute Plasmid Kit, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 350 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA purification: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Plasmid Miniprep Kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures. By combining silica-binding technology and the convenience of a spin column format, up to 15 μg of high copy plasmid DNA can be recovered from 1-5 mL of E. coli culture in less than 30 minutes. Note that actual yield and optimal volume of culture to use depend on the plasmid and the culture medium.

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the DNA onto silica in the presence of high salts. Contaminants are then removed by a spin-wash step. Finally, the bound DNA is eluted in water or Tris-EDTA buffer.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The DNA is ready for immediate use in applications such as restriction enzyme digestion, cloning, PCR, transformation, transcription, conventional, and automated sequencing.
The kit has been used for the purification of plasmid DNA of Dirofilaria repens.

Features and Benefits

  • Purified plasmid DNA in less than 30 minutes
  • Purify up to 15 μg of plasmid DNA
  • No detectable genomic DNA or RNA contamination
  • No phenol/chloroform extraction or alcohol precipitation required

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirofilaria repens: emergence of autochthonous human infections in the Czech Republic (case reports).
Mateju J
BMC Infectious Diseases, 16, 171-171 (2016)
Simeon S Andrews et al.
The Journal of biological chemistry, 294(30), 11549-11558 (2019-06-12)
Two-hybrid systems can be used for investigating protein-protein interactions and may provide important information about gene products with unknown function. Despite their success in mapping protein interactions, two-hybrid systems have remained mostly untouched by improvements in next-generation DNA sequencing. The
Mahmood Faraz et al.
The Journal of biological chemistry, 293(9), 3421-3435 (2018-01-11)
Leucine-rich repeats and immunoglobulin-like domains 1 (LRIG1) is a tumor suppressor and a negative regulator of several receptor tyrosine kinases. The molecular mechanisms by which LRIG1 mediates its tumor suppressor effects and regulates receptor tyrosine kinases remain incompletely understood. Here
Vanessa Meier-Stephenson et al.
The Journal of biological chemistry, 296, 100589-100589 (2021-03-29)
Approximately 250 million people worldwide are chronically infected with the hepatitis B virus (HBV) and are at increased risk of developing cirrhosis and hepatocellular carcinoma. The HBV genome persists as covalently closed circular DNA (cccDNA), which serves as the template
Jing Yang et al.
Preparative biochemistry & biotechnology, 50(6), 529-537 (2020-01-10)
As a lincosamide antibiotic, lincomycin is still important for treating diseases caused by Gram-positive bacteria. Manufacturing of lincomycin needs efforts to, e.g. maximize desirable species and minimizing unwanted fermentation byproducts. Analysis of the lincomycin biosynthetic gene cluster of Streptomyces lincolnensis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service