Skip to Content
Merck
All Photos(2)

Documents

M1196

Sigma-Aldrich

Anti-Mucolipin-2 (C-terminal) antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MCOLN2, Anti-MLN2, Anti-TRPML2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 0.3-0.6 μg/mL using HEK-293T cells expressing human mucolipin-2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCOLN2(255231)

General description

Mucolipin-2 (MLN2) is a member of transient receptor potential (TRP) protein superfamily. It consists of gated, tetrameric cation channels. MLN2 has a conserved structure of six transmembrane helices with differing cytoplasmic oriented N- and C-terminal domains. It is localized to late endosomes and lysosomes..

Specificity

Additional bands may be observed at ~120 kDa and ~35 kDa due to aggregation and degradation, respectively.

Immunogen

synthetic peptide corresponding to amino acids 534-549 of human mucolipin-2. This sequence has low homology with rat (73% identity) and mouse (66% homology) mucolipin-2.

Application

Anti-Mucolipin-2 (C-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Mucolipin-2 (MLN2) plays an important role in lysosome maturation, lysosome fission, late endosome-lysosome fusion, autophagosome-lysosome fusion, lysosome expulsion and iron release. It stimulates autophagy proliferation via transcription factor EB (TFEB) activation. MLN2 is involved in B‐lymphocyte development.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mucolipin-2 Cation Channel increases trafficking efficiency of endocytosed viruses
Rinkenberger N and Schoggins JW
mBio, 9(1), e02314-e02317 (2018)
Claudia Karacsonyi et al.
Traffic (Copenhagen, Denmark), 8(10), 1404-1414 (2007-07-31)
In mammals, the mucolipin family includes three members mucolipin-1, mucolipin-2, and mucolipin-3 (MCOLN1-3). While mutations in MCOLN1 and MCOLN3 have been associated with mucolipidosis type IV and the varitint-waddler mouse phenotype, respectively, little is known about the function and cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service