Skip to Content
Merck
All Photos(2)

Documents

I5286

Sigma-Aldrich

IL-3 from mouse

Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture

Synonym(s):

HCGF P-cell stimulation factorIL-3 MGC79398, Interleukin-3, MCGF (Mast cell growth factor)Multi-CSF, MGC79399

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

15.1 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... IL3(16187)

Looking for similar products? Visit Product Comparison Guide

General description

IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoieses, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is species-specific, variably glycosylated cytokine. Recombinant murine IL-3 is a 15.1 kDa globular protein containing 135 amino acid residues.

Biochem/physiol Actions

IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Recombinant murine IL-3 is a 15.1 kDa globular protein containing 135 amino acid residues.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadja Bakocevic et al.
European journal of immunology, 44(6), 1823-1834 (2014-03-13)
Basophils, a rare leukocyte population in peripheral circulation, are conventionally identified as CD45(int) CD49b(+) FcεRI(+) cells. Here, we show that basophils from blood and several organs of naïve wild-type mice express CD41, the α subunit of α(IIb)β₃ integrin. CD41 expression
Jing Wang et al.
EMBO molecular medicine, 6(7), 952-969 (2014-06-26)
Immunoglobulin E (IgE) activates mast cells (MCs). It remains unknown whether IgE also activates other inflammatory cells, and contributes to the pathogenesis of abdominal aortic aneurysms (AAAs). This study demonstrates that CD4+ T cells express IgE receptor FcεR1, at much
Serena De Vita et al.
PloS one, 9(5), e96209-e96209 (2014-05-03)
Systemic Mastocytosis (SM) is a clonal disease characterized by abnormal accumulation of mast cells in multiple organs. Clinical presentations of the disease vary widely from indolent to aggressive forms, and to the exceedingly rare mast cell leukemia. Current treatment of
Sofia von Palffy et al.
Experimental hematology, 43(7), 514-523 (2015-05-09)
Inactivating mutations in IKZF1, the gene that encodes the transcription factor IKAROS, are recurrent in poor-prognosis human B-cell leukemias, in which these mutations co-exist with BCR-ABL1 or other genetic changes that activate similar intracellular signaling pathways. However, little is known
Kerstin Kapp et al.
Molecular therapy. Nucleic acids, 3, e170-e170 (2014-06-25)
Toll-like receptors are sensing modulators of the innate immune system. One member of this protein family, Toll-like receptor (TLR)-9, is increasingly being investigated as therapeutic target for infectious diseases and cancer. Double-Stem Loop ImmunoModulator (dSLIM) is a new TLR-9 agonist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service