Skip to Content
Merck
All Photos(2)

Key Documents

HPA006303

Sigma-Aldrich

Anti-MYT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MTF1, Anti-MYTI, Anti-NZF2, Anti-PLPB1, Anti-ZC2HC4A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EAAPDVIFQEDTSHTSAQKAPELRGPESPSPKPEYSVIVEVRSDDDKDEDTHSRKSTVTDESEMQDMMTRGNLGLLEQAIALKAEQVRTVCEPGCPPAEQSQLGLGEPGKAAKPLDT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYT1(4661)

General description

Myelin transcription factor 1 is a protein encoded by the MYT1 gene in humans. It is a zinc-finger, DNA-binding protein and is known to be expressed in early progenitors of oligodendrocytes. It may be the initial member of a new structural family of zinc-finger DNA binding proteins.

Immunogen

Myelin transcription factor 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-MYT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

MYT1 is highly expressed in the developing nervous system, where expression is followed by the activation of its supposed target, proteolipid protein (PLP). It is suppressed by an M phase-activated kinase and inhibits mitosis by interacting with Cdc2. This gene may be useful as a target for anti-cancer therapies. In periventricular leukomalacia (PVL) regions, it might be involved in the repair of myelin. It might be involved in the assembly of transcriptionally active complexes of PLP.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86918

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay Chandrasekar et al.
Frontiers in behavioral neuroscience, 4, 14-14 (2010-04-22)
Brain-specific neural-zinc-finger transcription factor-2b (NZF2b/7ZFMyt1) is induced in the mesolimbic dopaminergic region after chronic cocaine exposure and lentiviral-mediated expression of NZF2b/7ZFMyt1 in the nucleus accumbens results in decreased locomotor activity (Chandrasekar and Dreyer, 2010). In this study the role of
Vijay Chandrasekar et al.
Neurobiology of disease, 37(1), 86-98 (2009-09-30)
Chronic cocaine induces high expression of the brain-specific Neural-Zinc-Finger transcription factor-2b (NZF-2b/7ZFMyt1), particularly in the mesolimbic dopaminergic pathway, resulting in a 11-fold increase in NZF-2b/7ZFMyt1 expression in the Nucleus Accumbens (NAc). Overexpression of this gene in the NAc with a
Adam C Vana et al.
Glia, 55(7), 687-697 (2007-03-03)
Myelin transcription factor 1 (Myt1) is a zinc-finger DNA binding protein that influences developing oligodendrocyte progenitor (OP) cell proliferation, differentiation, and myelin gene transcription in vitro. The potential of Myt1 to play a role in OP responses leading to remyelination
R N Booher et al.
The Journal of biological chemistry, 272(35), 22300-22306 (1997-08-29)
Activation of the Cdc2.cyclin B kinase is a pivotal step of mitotic initiation. This step is mediated principally by the dephosphorylation of residues threonine 14 (Thr14) and tyrosine 15 (Tyr15) on the Cdc2 catalytic subunit. In several organisms homologs of
J P H Chow et al.
Oncogene, 32(40), 4778-4788 (2012-11-14)
Inhibition of cyclin-dependent kinase 1 (CDK1) by phosphorylation is a key regulatory mechanism for both the unperturbed cell cycle and the DNA damage checkpoint. Although both WEE1 and MYT1 can phosphorylate CDK1, little is known about the contribution of MYT1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service