Skip to Content
Merck
All Photos(7)

Key Documents

HPA003363

Sigma-Aldrich

Anti-FBXO44 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-F-box only protein 44 antibody produced in rabbit, Anti-F-box protein FBX30 antibody produced in rabbit, Anti-F-box/G-domain protein 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
7
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SLHRNLLHNPCAEEGFEFWSLDVNGGDEWKVEDLSRDQRKEFPNDQVRSQARLRVQVPAVRSAPVVRARASGDLPARPGDHPAEERCQVEGGLPHILQLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXO44(93611)

Immunogen

F-box only protein 44 recombinant protein epitope signature tag (PrEST)

Application

Anti-FBXO44 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FBXO44 (F-box protein 44) gene encodes a protein belonging to the F-box protein family. It contains a conserved G domain that is also called as FBA domain. This is a sugar recognition domain that mediates substrate binding for BRCA1 ubiquitination and subsequent degradation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86596

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Carlucci et al.
British journal of cancer, 112(1), 9-13 (2014-12-10)
Mutations in the BRCA1 and BRCA2 genes predispose individuals to the development of breast and ovarian cancers. As a result, biochemical functions of BRCA1 and BRCA2 proteins are being characterised in great detail. These studies have prompted the use of
Yunzhe Lu et al.
The Journal of biological chemistry, 287(49), 41014-41022 (2012-10-23)
BRCA1 mutations account for a significant proportion of familial breast and ovarian cancers. In addition, reduced BRCA1 protein is associated with sporadic cancer cases in these tissues. At the cellular level, BRCA1 plays a critical role in multiple cellular functions
Rebecca R Florke Gee et al.
Acta pharmaceutica Sinica. B, 13(11), 4523-4534 (2023-11-16)
Pregnane X receptor (PXR) is a ligand-activated nuclear receptor that transcriptionally upregulates drug-metabolizing enzymes [e.g., cytochrome P450 3A4 (CYP3A4)] and transporters. Although the regulation of PXR target genes is well-characterized, less is known about the regulation of PXR protein level.
Kevin A Glenn et al.
The Journal of biological chemistry, 283(19), 12717-12729 (2008-01-22)
Post-translational modification of proteins regulates many cellular processes. Some modifications, including N-linked glycosylation, serve multiple functions. For example, the attachment of N-linked glycans to nascent proteins in the endoplasmic reticulum facilitates proper folding, whereas retention of high mannose glycans on
Harrison J McNabb et al.
Molecular pharmacology, 98(6), 677-685 (2020-10-04)
Regulator of G protein signaling (RGS) proteins are negative modulators of G protein signaling that have emerged as promising drug targets to improve specificity and reduce side effects of G protein-coupled receptor-related therapies. Several small molecule RGS protein inhibitors have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service