Skip to Content
Merck
All Photos(2)

Key Documents

HPA002727

Sigma-Aldrich

Anti-NRXN3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neurexin 3 isoform α precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

LTIFNTQAQIAIGGKDKGRLFQGQLSGLYYDGLKVLNMAAENNPNIKINGSVRLVGEVPSILGTTQTTSMPPEMSTTVMETTTTMATTTTRKNRSTASIQPTSDDLVSSAECSSDDEDFVECEPSTANPTEP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRXN3(9369)

General description

NRXN3 (neurexin 3) is a neuronal cell surface protein, also expressed in the lung, pancreas, heart, placenta, liver and kidney. In humans, three types of neurexin genes are present i.e. NRXN1, NRXN2, and NRXN3. It belongs to the family of polymorphic cell-surface proteins.

Immunogen

Neurexin 3 isoform α precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NRXN3 (neurexin 3) is mainly involved in the intercellular signaling pathways and neuronal cell adhesion during synaptogenesis. It participates in different intercellular connections with the help of extracellular matrix protein and other proteins. The gene may play a role in schizophrenia, autism and obesity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70533

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Hu et al.
Progress in neuro-psychopharmacology & biological psychiatry, 43, 197-202 (2013-01-12)
Recent researches have implicated that mutations in the neurexin-3 (NRXN3) gene on chromosome 14q24.3-q31.1 might play a role in addiction, autism, and obesity. In order to explore the association of NRXN3 polymorphisms with schizophrenia, we examined seven single nucleotide polymorphisms
M Missler et al.
Trends in genetics : TIG, 14(1), 20-26 (1998-02-04)
The human brain has approximately 10(12) neurons, three orders of magnitude more than there are basepairs in the human genome. Each neuron is connected to other neurons by thousands of synapses, creating a dense network of communicating neurons. Cell-recognition events
Gianluca Occhi et al.
Biochemical and biophysical research communications, 298(1), 151-155 (2002-10-16)
Three neurexin (NRXN) genes are known in humans, each transcribed from two promoters and extensively spliced at five canonical positions, thus generating thousands of isoforms. For NRXN3, only neuronal expression was reported so far. We reported here on the expression
Lee Rowen et al.
Genomics, 79(4), 587-597 (2002-04-12)
The neurexins are neuronal proteins that function as cell adhesion molecules during synaptogenesis and in intercellular signaling. Although mammalian genomes contain only three neurexin genes, thousands of neurexin isoforms may be expressed through the use of two alternative promoters and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service