Skip to Content
Merck
All Photos(1)

Key Documents

GE28-4112-11

HITRAP® AVB SEPHAROSE HP

Cytiva 28-4112-11, pack of 5x1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 5x1 mL

manufacturer/tradename

Cytiva 28-4112-11

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

cross-linked agarose

avg. part. size

34 μm

cleaning

2-12

working range

3-10

capacity

1012 genecopies mL binding capacity

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

AVB Sepharose High performance is an affinity medium with affinity for adeno associated viruses. It enables purification of adeno associated viruses in one step with high purity and excellent yields. It has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5, but it is expected to have affinity for most of the other adeno associated virus subclasses. The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast.

Features and Benefits

  • One step purification of adeno associated viruses
  • Affinity medium with high selectivity and low background binding
  • Large surface area to give enhanced capacity
  • Prepacked HiTrap columns available, for simple operation with a syringe, pump, or chromatographic system

Storage and Stability

Store at 4 to 8 °C. (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service