Skip to Content
Merck
All Photos(2)

Key Documents

EP4960836001

Eppendorf® Bottle Thread Adapter

from GL 32 to GL 38, ETFE

Synonym(s):

Eppendorf® Bottle Thread Adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.15

material

ETFE

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4960836001

Looking for similar products? Visit Product Comparison Guide

Application

LIQUID HANDLING

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service