Skip to Content
Merck
All Photos(1)

Key Documents

EHU115211

Sigma-Aldrich

MISSION® esiRNA

targeting human MKNK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGACTTTGACTTGGGCAGTGGGATGAAACTGAACAACTCCTGTACCCCCATAACCACACCAGAGCTGACCACCCCATGTGGCTCTGCAGAATACATGGCCCCTGAGGTAGTGGAGGTCTTCACGGACCAGGCCACATTCTACGACAAGCGCTGTGACCTGTGGAGCCTGGGCGTGGTCCTCTACATCATGCTGAGTGGCTACCCACCCTTCGTGGGTCACTGCGGGGCCGACTGTGGCTGGGACCGGGGCGAGGTCTGCAGGGTGTGCCAGAACAAGCTGTTTGAAAGCATCCAGGAAGGCAAGTATGAGTTTCCTGACAAGGACTGGGCACACATCTCCAGTGAAGCCAAAGACCTCATCTCCAAGCTCCTGGTGCGAGATGCAAAGCAGAGACTTAGCGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C Brown et al.
Journal of virology, 88(22), 13149-13160 (2014-09-05)
Translation machinery is a major recipient of the principal mitogenic signaling networks involving Raf-ERK1/2 and phosphoinositol 3-kinase (PI3K)-mechanistic target of rapamycin (mTOR). Picornavirus internal ribosomal entry site (IRES)-mediated translation and cytopathogenic effects are susceptible to the status of such signaling
Sergio Rius-Pérez et al.
Scientific reports, 9(1), 3775-3775 (2019-03-09)
p38α MAPK negatively regulates the G1/S and G2/M cell cycle transitions. However, liver-specific p38α deficiency impairs cytokinesis and reduces hepatocyte proliferation during cirrhosis and aging in mice. In this work, we have studied how p38α down-regulation affects hepatocyte proliferation after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service