E6287
Pectinase from Aspergillus aculeatus
Synonym(s):
Pectinex Ultra Clear®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Aspergillus aculeatus
Quality Level
form
aqueous solution
concentration
≥800 units/mL
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Pectinase from Aspergillus aculeatus is used in plant protoplast preparation to digest cell wall prior to organelle isolation. It has been used to conduct partial saccharification of sugars. Pectinases are used to study their role in the invasion of plant tissues using phytopathogens, as well as various food processing and plant biotechnology applications. The enzyme from Sigma has been used to determine the content of quercetin produced and also to evaluate its rutinase activity.
Biochem/physiol Actions
Pectinase is an active pectolytic enzyme preparation that is produced by a selected strain of Aspergillus aculeatus. It contains mainly pectintranseliminase, polygalacturonase and pectinesterase, along with small amounts of hemicellulases and cellulases. Pectinase hydrolyzes pectin, which is a component of the cell wall. They may attack methyl-esterified pectin or de-esterified pectin.
Other Notes
View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer
Legal Information
A product of Novozyme Corp.
Pectinex is a registered trademark of Novozymes Corp.
UltraClear is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ting Sun et al.
Journal of agricultural and food chemistry, 53(1), 42-48 (2005-01-06)
Commercial pectolytic enzymes were investigated for their influence on phenolics and antioxidant activities of asparagus juice. The antioxidant activity of asparagus juice was analyzed according to 2,2'-diphenyl-l-picrylhydrazyl and 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonic acid) methods. The enzymes, with the exception of pectinase from Rhizopus
Dariusz J Smoliński et al.
Histochemistry and cell biology, 136(5), 527-541 (2011-09-10)
Small nuclear ribonucleoproteins (snRNPs) play a fundamental role in pre-mRNA processing in the nucleus. The biogenesis of snRNPs involves a sequence of events that occurs in both the nucleus and cytoplasm. Despite the wealth of biochemical information about the cytoplasmic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service