CLS353143
Corning® Falcon® Cell Culture Multi-Flask
capacity 525 mL, 3-layer, cap, vented
Synonym(s):
Corning container
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
Recommended Products
material
polystyrene
feature
cap vented
manufacturer/tradename
Corning 353143
technique(s)
cell culture | mammalian: suitable
capacity
525 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service