CLS11495
Corning® Erlenmeyer flask, with aseptic connector
volume 3 L, I.D. 1/4 in., sterile
Synonym(s):
Corning container
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
Recommended Products
material
flat bottom flask
polycarbonate
polypropylene cap
description
plain
sterility
sterile
feature
cap: no
C-Flex tubing
packaging
case of 2
manufacturer/tradename
Corning 11495
I.D.
1/4 in.
capacity
3 L
volume
3 L
3000 mL
fitting
male luer lock
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service