Skip to Content
Merck
All Photos(2)

Key Documents

CLL1031

Sigma-Aldrich

U2OS GFP-TUBA1B

human female bone (Disease source: Osteosarcoma)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

Product Name

U2OS GFP-TUBA1B,

biological source

human female bone (Source Disease: Osteosarcoma)

OMIM accession no.

storage temp.

−196°C

Gene Information

General description

K-α1 tubulin/ tubulin α 1b (TUBA1B) is a microtubule protein. It is a ~50 kDa protein in its poly glycosylated, post-translational form. TUBA1B is present in cells as one of the six different isotypes. This gene is located on human chromosome 12q13.12.
This product corresponds to ATCC Cat. No. HTB-96.
U2OS GFP-TUBA1B are osteosarcoma, epithelial cells, from a human caucasian female (aged 15 years), with a GFP-tagged modification.

Application

This product is a human U2OS cell line in which the genomic TUBA1B gene has been endogenously tagged with a Green Fluorescent Protein (GFP) gene using CompoZr® Zinc Finger Nuclease technology. Integration resulted in endogenous expression of the fusion protein in which GFP is attached to the N-terminus of tubulin. Fluorescence imaging shows that the tubulin can polymerize to form the characteristic pattern of microtubules. Addition of vincristine to this cell line shows the expected depolymerization of microtubules. This stable cell line was expanded from a single clone. The target′s gene regulation and corresponding protein function are preserved in contrast to cell lines with overexpression via an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage

Biochem/physiol Actions

K-α1 tubulin/ tubulin α 1b (TUBA1B) participates in several cellular activities like GTP binding, GTPase activity, maintaining cellular structure in the form of microtubules and microtubule-based intracellular movement. It plays a crucial role in polymerization and depolymerization during cell replication and division. TUBA1B is overexpressed in hepatocellular carcinoma (HCC) tumor tissues and proliferating HCC cells.
Tubulin α 1b (TUBA1B) plays an important role in dynamic process of polymerization and depolymerization during cell replication and division. Elevated expression of the gene has been observed in hepatocellular carcinoma (HCC) tumor tissues and proliferating HCC cells. This upregulated expression of the gene leads to poor prognosis and chemotherapy resistance in HCC.

Features and Benefits

GFP-tagged on chromosome 12q13.12
The U2OS cells are adherent, with a doubling time of approx. 29 hours.

Components

U2OS GFP-TUBA1B cells

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Preparation Note

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:6

The base medium for this cell line is McCoy′s 5A1 Medium Modified, Cat. No. M9309. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration of 10%.

The cell freezing medium is DMSO 1X (Cat. No. C6164).

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo production of K-α1 tubulin-specific antibodies: role in chronic lung allograft rejection.
Goers TA, et al.
Journal of Immunology, 180(7), 4487-4494 (2008)
Up-regulation of TUBA1B promotes astrocyte proliferation after spinal cord injury in adult rats.
Liu Y, et al.
International Journal of Clinical and Experimental Pathology, 10(2), 1094-1103 (2017)
Increased α-tubulin1b expression indicates poor prognosis and resistance to chemotherapy in hepatocellular carcinoma.
Lu C, et al.
Digestive Diseases and Sciences, 58(9), 2713-2720 (2013)
A revised nomenclature for the human and rodent alpha-tubulin gene family.
Khodiyar VK
Genomics, 90(2), 285-289 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service