Skip to Content
Merck
All Photos(1)

Key Documents

HEPM

90120505, human palate epithelium. Established from palatal shelves.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HEPM, 90120505

biological source

human mouth (embryonic)

packaging

tube of 5 μg 90120505-DNA-5UG
pkg of vial of cells 90120505-1VL

growth mode

Adherent

karyotype

2n = 46, diploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human embryonic palatal mesenchyme

Cell Line Description

Established from embryonic palatal shelves. The cells are very sensitive to the stimulating effect of EGF with respect to proliferation, incorporation of thymidine and ornithine decarboxylase.

Application

Proliferation studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service