Skip to Content
Merck
All Photos(1)

Documents

H4II

89042702, rat skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

H4II, 89042702

biological source

rat skin

growth mode

Adherent

karyotype

52 chromosomes

morphology

Epithelial

products

Albumin, transferrin, prothrombin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat skin Hepatocellular carcinoma

Cell Line Description

From H-35 Reuber hepatoma induced by chemical carcinogenesis in an AxC rat . Tyrosine aminotransferase positive. Synthesises several serum proteins including albumin, transferrin and prothrombin. (Same as H4-11-E-C3 (Sigma Catalogue number. 85061112)).

Application

Gene expression analysis.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent [60%-70%] cultures 1:100 i.e. seeding at 2x100 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density is reached at 15x1000,000 cells/10cm2 dish.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service