Skip to Content
Merck
All Photos(1)

Key Documents

HT 1376

87032402, human bladder, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HT 1376, 87032402

biological source

human bladder

growth mode

Adherent

karyotype

2n = 46, hypertetrapoid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian bladder carcinoma

Cell Line Description

Derived from a 58 year old caucasian female with a grade 3 transitional carcinoma of the bladder who had not received chemo or radiation therapy. The cells grow in soft agar and are tumorigenic in mice and hamsters. The patient donor had not received chemotherapy or radiation treatment.

Application

Tumourigenicity studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service