Skip to Content
Merck
All Photos(2)

Key Documents

C4124

Sigma-Aldrich

Chromium picolinate

≥98% (HPLC)

Synonym(s):

Cr(pic)3, CrPic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12CrN3O6
CAS Number:
Molecular Weight:
418.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98% (HPLC)

form

powder

loss

≤5% loss on drying

storage temp.

room temp

SMILES string

O=C(O[Cr](OC(=O)c1ccccn1)OC(=O)c2ccccn2)c3ccccn3

InChI

1S/3C6H5NO2.Cr/c3*8-6(9)5-3-1-2-4-7-5;/h3*1-4H,(H,8,9);/q;;;+3/p-3

InChI key

CBDQOLKNTOMMTL-UHFFFAOYSA-K

General description

Chromium picolinate is a supplemental or alternative medication for type 2 diabetes and an anti-diabetic mechanisms of CrPic in insulin-resistant 3T3-L1 adipocytes.

Application

Chromium picolinate (CrPic), a dietay supplement, may be used to study its potential as a modulator of glucose uptake and the activity of insulin. CrPic is used to study its effects on nuclear factor-κ B (NF-κB) and nuclear factor-E2-related factor-2 (Nrf2) pathways in animals.

Biochem/physiol Actions

Chromium picolinate (CrPic) is a supplemental or alternative medication for type 2 diabetes. Experimental evidence shows the effect of CrPic on glucose uptake through the activation of p38 MAPK. Chromium is thought to potentiate the action of insulin, leading to increased insulin sensitivity in type 2 diabetes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Appleton et al.
Journal of feline medicine and surgery, 4(1), 13-25 (2002-03-01)
The effect of dietary chromium supplementation on glucose and insulin metabolism in healthy, non-obese cats was evaluated. Thirty-two cats were randomly divided into four groups and fed experimental diets consisting of a standard diet with 0 ppb (control), 150 ppb
C L Depew et al.
Journal of dairy science, 81(11), 2916-2923 (1998-12-05)
Forty-two Holstein calves were used to study performance and metabolic responses when milk replacer and then postweaning starter were supplemented with 1 ppm of Cr as Cr-tripicolinate. From birth through 8 wk of age, supplemental Cr tended to improve the
L R Gentry et al.
Journal of animal science, 77(5), 1284-1294 (1999-05-26)
Thirty-two Suffolk wether lambs were fed for 84 d in a 2 x 2 factorial experiment using two levels of dietary protein (9.0 to 12.1% CP, low protein, LP; or 12.8 to 14.4% CP, high protein, HP) and supplemental Cr
M D Lindemann et al.
Journal of animal science, 82(10), 2972-2977 (2004-10-16)
A cooperative research study involving 353 litters was conducted at three stations to determine the effects of graded levels of supplemental Cr from chromium tripicolinate (CrPic) on reproductive performance of sows and preweaning performance of their pigs. Primiparous and multiparous
T L Ward et al.
Journal of animal science, 75(4), 1001-1008 (1997-04-01)
An experiment was conduct to determine the interactive effects of dietary chromium tripicolinate (CrPic), CP, and pen space (PS) as a stress on growth performance, carcass traits, and plasma hormones and metabolites of crossbred growing-finishing pigs (128 pigs; initial BW

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service