AXYPCR02C
PCR tubes with flat caps
thin wall, size 0.2 mL, pkg of 10x1000tubes/cs, clear
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
Recommended Products
material
cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-02-C
Corning
packaging
pkg of 10x1000tubes/cs
technique(s)
PCR: suitable
size
0.2 mL
color
clear
fitting
cap (push fit)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Manufactured with 99.9% pure virgin polypropylene.
- Thin-wall PCR tubes with flat caps are designed to fit standard 96 well blocks.
- Ultra-thin and consistent wall thickness allow the precise thermal transfer.
- Even insertion cap ensures a tight fit to prevent evaporation.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service