Skip to Content
Merck
All Photos(4)

Documents

AV100621

Sigma-Aldrich

Anti-SMAD3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-SMAD family member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

dog, rat, human, mouse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD3(4088)

General description

The members of Smad family of proteins are essential intracellular mediators of TGF-β signaling.

Immunogen

Synthetic peptide directed towards the N terminal region of human SMAD3

Application

Anti-SMAD3 antibody produced in rabbit is suitable for western blotting at a concentration of 2 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

Smad3 is phosphorylated by TGF-b, heterodimerizes with Smad4 and enters the nucleus. The heterodimer binds within the promoter of plasminogen activator inhibitor-1 (PAI-1) and induces its expression.

Sequence

Synthetic peptide located within the following region: FTPPIVKRLLGWKKGEQNGQEEKWCEKAVKSLVKKLKKTGQLDELEKAIT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Nakao et al.
The EMBO journal, 16(17), 5353-5362 (1997-10-06)
Smad family members are newly identified essential intracellular signalling components of the transforming growth factor-beta (TGF-beta) superfamily. Smad2 and Smad3 are structurally highly similar and mediate TGF-beta signals. Smad4 is distantly related to Smads 2 and 3, and forms a
Yuze Zhang et al.
Cardiovascular diabetology, 20(1), 121-121 (2021-06-13)
Cardiac remodeling is one of the major risk factors for heart failure. In patients with type 2 diabetes, sodium-glucose cotransporter 2 (SGLT2) inhibitors reduce the risk of the first hospitalization for heart failure, possibly through glucose-independent mechanisms in part, but
L Zawel et al.
Molecular cell, 1(4), 611-617 (1998-07-14)
Mounting evidence indicates that Smad proteins are required for TGF beta signaling, but the way(s) in which Smad proteins propagate this signal is unclear. We found that two human Smad proteins (Smad3 and Smad4) could specifically recognize an identical 8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service