Skip to Content
Merck
All Photos(1)

Key Documents

A7785

Sigma-Aldrich

Apolipoprotein C-I from human plasma

≥95% (SDS-PAGE), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

average mol wt 6.6 kDa

technique(s)

inhibition assay: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APOC1(341)

General description

Very low density lipoprotein.

Application

Apolipoprotein C-1 directly interferes with fatty acid uptake and is a major plasma inhibitor of cholesterol ester transfer protein. Apolipoprotein C-I has been used to develop a quantitative strategy, named secretome-derived isotopic tag (SDIT), to concurrently identify and quantify the adipocyte-secreted plasma proteins from normal and high-fat-diet (HFD) induced obese mice.

Biochem/physiol Actions

Interferes directly with fatty acid uptake and is the major plasma inhibitor of cholesterol ester transfer protein.

Physical form

Lyophilized from 10 mM NH4HCO3, pH 7.5

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B S Suri et al.
The Biochemical journal, 178(2), 455-466 (1979-02-15)
1. The work reported was designed to provide quantitative information about the capacity of the extrahepatic tissues of the rat to degrade injected VLD lipoproteins (very-low-density lipoproteins, d less than 1.006) to LD lipoproteins (low-density lipoproteins, d 1.006--1.063) and to
Rong-Xia Li et al.
Journal of proteome research, 11(5), 2851-2862 (2012-03-13)
We developed a quantitative strategy, named secretome-derived isotopic tag (SDIT), to concurrently identify and quantify the adipocyte-secreted plasma proteins from normal and high-fat-diet (HFD) induced obese mice, based on the application of isotope-labeled secreted proteins from cultured mouse adipocytes as
Nathan L Meyers et al.
Biochemistry, 51(6), 1238-1248 (2012-01-24)
Apolipoprotein C-I (apoC-I) is an important constituent of high-density lipoprotein (HDL) and is involved in the accumulation of cholesterol ester in nascent HDL via inhibition of cholesterol ester transfer protein and potential activation of lecithin:cholesterol acyltransferase (LCAT). As the smallest

Articles

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service