Skip to Content
Merck
All Photos(1)

Key Documents

54747

Sigma-Aldrich

γ,γ-Dimethylallyl phosphate ammonium salt

≥93.0% (TLC)

Synonym(s):

3,3-Dimethylallyl phosphate ammonium salt, 3-Methyl-2-butenyl phosphate ammonium salt, Phosphoric acid mono-(3-methyl-2-butenyl ester) ammonium salt, Prenyl phosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11O4P · xNH3
Molecular Weight:
166.11 (free acid basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥93.0% (TLC)

storage temp.

−20°C

SMILES string

N.C\C(C)=C\COP(O)(O)=O

InChI

1S/C5H11O4P.H3N/c1-5(2)3-4-9-10(6,7)8;/h3H,4H2,1-2H3,(H2,6,7,8);1H3

InChI key

FLEGYDAACQHRAY-UHFFFAOYSA-N

Application

Gamma, gamma-dimethylallyl phosphate may be useful in studies of the archaeal isopentenyl phosphate kinases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mo Chen et al.
Biochemistry, 49(1), 207-217 (2009-11-26)
Archaea synthesize isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP), the essential building blocks of isoprenoid compounds, from mevalonate (MVA). However, an analysis of the genomes of several members of the Archaea failed to identify genes for the enzymes required to
Nikki Dellas et al.
ACS chemical biology, 5(6), 589-601 (2010-04-16)
The biosynthesis of isopentenyl diphosphate (IPP) from either the mevalonate (MVA) or the 1-deoxy-d-xylulose 5-phosphate (DXP) pathway provides the key metabolite for primary and secondary isoprenoid biosynthesis. Isoprenoid metabolism plays crucial roles in membrane stability, steroid biosynthesis, vitamin production, protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service